Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6,7,8-trifluoro-4-oxohydroquinoline-3-carboxylate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
79660-46-1
C12H8F3NO3
271.195
MFCD00276021
ONQDAESGZUODFI-UHFFFAOYSA-N
ethyl 6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate,dm-7,ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate,3-quinolinecarboxylic acid, 1,4-dihydro-4-oxo-6,7,8-trifluoro-, ethyl ester,ethyl 6,7,8-trifluoro-4-hydroxyquinoline-3-carboxylate,ethyl 1,4-dihydro-4-oxo-6,7,8-trifluoro-3-quinolinecarboxylate,ethyl 6,7,8-trifluoro-4-oxohydroquinoline-3-carboxylate,6,7,8-trifluoro-4-hydroxyquinoline-3-carboxylicacidethylester,ethyl 4-hydroxy-6,7,8-trifluoroquinoline-3-carboxylate
2733972
ethyl 6,7,8-trifluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C(=C(C=C2C1=O)F)F)F
79660-46-1
C12H8F3NO3
271.195
MFCD00276021
ONQDAESGZUODFI-UHFFFAOYSA-N
ethyl 6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate,dm-7,ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate,3-quinolinecarboxylic acid, 1,4-dihydro-4-oxo-6,7,8-trifluoro-, ethyl ester,ethyl 6,7,8-trifluoro-4-hydroxyquinoline-3-carboxylate,ethyl 1,4-dihydro-4-oxo-6,7,8-trifluoro-3-quinolinecarboxylate,ethyl 6,7,8-trifluoro-4-oxohydroquinoline-3-carboxylate,6,7,8-trifluoro-4-hydroxyquinoline-3-carboxylicacidethylester,ethyl 4-hydroxy-6,7,8-trifluoroquinoline-3-carboxylate
2733972
ethyl 6,7,8-trifluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C(=C(C=C2C1=O)F)F)F
Packaging Glass Bottle
Melting Point 267°C to 272°C
Quantity 5g
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Formula Weight 271.2
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material Ethyl 6, 7, 8-trifluoro-4-oxohydroquinoline-3-carboxylate

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RTECSNumber : VB2009830