Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 6-amino-5-methoxyindole-2-carboxylate, 95%

Click to view available options
Quantity:
1g
5g
107575-60-0
C12H14N2O3
234.255
MFCD08064224
DIEUGINJYISDCB-UHFFFAOYSA-N
ethyl 6-amino-5-methoxyindole-2-carboxylate,acmc-209zno,ethyl6-amino-5-methoxy-1h-indole-2-carboxylate,ethyl 6-amino-5-methoxy-1h-indole-2-carboxylate,1h-indole-2-carboxylicacid, 6-amino-5-methoxy-, ethyl ester
10966428
ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC(=C(C=C2N1)N)OC
107575-60-0
C12H14N2O3
234.255
MFCD08064224
DIEUGINJYISDCB-UHFFFAOYSA-N
ethyl 6-amino-5-methoxyindole-2-carboxylate,acmc-209zno,ethyl6-amino-5-methoxy-1h-indole-2-carboxylate,ethyl 6-amino-5-methoxy-1h-indole-2-carboxylate,1h-indole-2-carboxylicacid, 6-amino-5-methoxy-, ethyl ester
10966428
ethyl 6-amino-5-methoxy-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=CC(=C(C=C2N1)N)OC
Applications
Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Quantity 1g
Melting Point 167°C to 169°C
Solubility Information Soluble in dichloromethane,ethyl acetate and methanol.
Formula Weight 234.25
Percent Purity 95%
Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures