Alfa Aesar™ Ethyl 6-aminohexanoate, 98%

Manufacturer: Alfa Aesar™ H50296MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50296MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 371-34-6
    Molecular Formula C8H17NO2
    Linear Formula H2N(CH2)5CO2CH2CH3
    Formula Weight 159.23
    MDL Number MFCD00233495
    Synonym 6-Aminocaproic acid ethyl ester; 6-Aminohexanoic acid ethyl ester
    Chemical Name or Material Ethyl 6-aminohexanoate

    Specifications

    Formula Weight 159.23
    Molecular Formula C8H17NO2
    Linear Formula H2N(CH2)5CO2CH2CH3
    MDL Number MFCD00233495
    Synonym 6-Aminocaproic acid ethyl ester; 6-Aminohexanoic acid ethyl ester
    CAS 371-34-6
    Quantity 250mg
    Chemical Name or Material Ethyl 6-aminohexanoate
    Assay 98%

    Description

    Applications
    Ethyl 6-aminohexanoate is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit