Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate, 98%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
59128-04-0
C11H11BrN2O2
283.125
MFCD16172180
VBLCCEAJXYZCRL-UHFFFAOYSA-N
ethyl 2-6-bromoimidazo 1,2-a pyridin-2-yl acetate,ethyl 2-6-bromoimidazo 1,2-a pyridin-2-yl acetate,ethyl 6-bromoimidazo 1,2-a pyridine-2-yl acetate,6-bromo-imidazo 1,2-a pyridin-2-yl-acetic acid ethyl ester,imidazo 1,2-a pyridine-2-acetic acid, 6-bromo-, ethyl ester
12269134
ethyl 2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
CCOC(=O)CC1=CN2C=C(C=CC2=N1)Br
59128-04-0
C11H11BrN2O2
283.125
MFCD16172180
VBLCCEAJXYZCRL-UHFFFAOYSA-N
ethyl 2-6-bromoimidazo 1,2-a pyridin-2-yl acetate,ethyl 2-6-bromoimidazo 1,2-a pyridin-2-yl acetate,ethyl 6-bromoimidazo 1,2-a pyridine-2-yl acetate,6-bromo-imidazo 1,2-a pyridin-2-yl-acetic acid ethyl ester,imidazo 1,2-a pyridine-2-acetic acid, 6-bromo-, ethyl ester
12269134
ethyl 2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
CCOC(=O)CC1=CN2C=C(C=CC2=N1)Br
Melting Point 55°C to 56°C
Quantity 250mg
Color Gray
Beilstein 791271
Formula Weight 283.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning