Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 6-chloroindole-2-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
27034-51-1
C11H10ClNO2
223.656
MFCD03084731
FSMZLIBWSAMADK-UHFFFAOYSA-N
ethyl 6-chloroindole-2-carboxylate,6-chloroindole-2-carboxylic acid ethyl ester,6-chloro-1h-indole-2-carboxylic acid ethyl ester,6-chloro-2-indolecarboxylic methyl ester,1h-indole-2-carboxylic acid, 6-chloro-, ethyl ester,pubchem1672,ethyl 6-chloroindole-2 carboxylate,ethyl-6-chloroindole-2-carboxylate,ethyl 6-chloranyl-1h-indole-2-carboxylate,6-chlorindole-2-carboxylic acid ethyl ester
7015027
ethyl 6-chloro-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Cl
27034-51-1
C11H10ClNO2
223.656
MFCD03084731
FSMZLIBWSAMADK-UHFFFAOYSA-N
ethyl 6-chloroindole-2-carboxylate,6-chloroindole-2-carboxylic acid ethyl ester,6-chloro-1h-indole-2-carboxylic acid ethyl ester,6-chloro-2-indolecarboxylic methyl ester,1h-indole-2-carboxylic acid, 6-chloro-, ethyl ester,pubchem1672,ethyl 6-chloroindole-2 carboxylate,ethyl-6-chloroindole-2-carboxylate,ethyl 6-chloranyl-1h-indole-2-carboxylate,6-chlorindole-2-carboxylic acid ethyl ester
7015027
ethyl 6-chloro-1H-indole-2-carboxylate
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Cl
Quantity 1g
Beilstein 169178
Formula Weight 223.66
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloroindole-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning