Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, 97%, Maybridge

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Amber Glass Bottle
71083-06-2
C12H10FNO3
235.214
MFCD00052242
MPUYCZQHTGRPNE-UHFFFAOYSA-N
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate,ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate,ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate,3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester,8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester,8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester,3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester,pubchem19599,maybridge1_000540,acmc-209nc3
707160
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
71083-06-2
C12H10FNO3
235.214
MFCD00052242
MPUYCZQHTGRPNE-UHFFFAOYSA-N
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate,ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate,ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate,3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester,8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester,8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester,3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester,pubchem19599,maybridge1_000540,acmc-209nc3
707160
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
Packaging Amber Glass Bottle
Quantity 1g
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 235.21
Percent Purity 97%
Chemical Name or Material Ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.