Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl 9-fluorenylideneacetate, 98%

Click to view available options
Quantity:
10g
50g
27973-36-0
C17H14O2
250.297
MFCD00798562
SCHVJDVNASXDFU-UHFFFAOYSA-N
ethyl 9-fluorenylideneacetate,ethyl 2-9h-fluoren-9-ylidene acetate,ethyl 2-fluoren-9-ylidene acetate,acmc-1cei0,ethyl 9-fluorenylidene acetate,ethyl 9h-fluoren-9-ylideneacetate,ethyl 2-fluoren-9-ylideneethanoate,ethyl 9h-fluoren-9-ylideneacetate #,9-fluorenylideneacetic acid ethyl ester
282552
ethyl 2-fluoren-9-ylideneacetate
CCOC(=O)C=C1C2=CC=CC=C2C3=CC=CC=C31
27973-36-0
C17H14O2
250.297
MFCD00798562
SCHVJDVNASXDFU-UHFFFAOYSA-N
ethyl 9-fluorenylideneacetate,ethyl 2-9h-fluoren-9-ylidene acetate,ethyl 2-fluoren-9-ylidene acetate,acmc-1cei0,ethyl 9-fluorenylidene acetate,ethyl 9h-fluoren-9-ylideneacetate,ethyl 2-fluoren-9-ylideneethanoate,ethyl 9h-fluoren-9-ylideneacetate #,9-fluorenylideneacetic acid ethyl ester
282552
ethyl 2-fluoren-9-ylideneacetate
CCOC(=O)C=C1C2=CC=CC=C2C3=CC=CC=C31
Melting Point 75°C to 77°C
Quantity 10g
Formula Weight 250.3
Percent Purity 98%
Chemical Name or Material Ethyl 9-fluorenylideneacetate

TSCA : No

Recommended Storage : Ambient temperatures