Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

High quality chemicals for laboratory and specialized industrial use

Formula Weight 88.11
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.90kg
Flash Point −4.00°C
Quantity 4L
Chemical Name or Material Ethyl Acetate
Grade Baker Analyzed™ ACS Reagent