Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

High quality chemicals for laboratory and specialized industrial use

Formula Weight 88.11
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.90kg
Flash Point −4.00°C
Quantity 500mL
Chemical Name or Material Ethyl Acetate