Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate Ethanol Solution, 3:1 (v/v), Optima™ for HPLC, Fisher Chemical™

Click to view available options
Quantity:
1L
4L
141-78-6, 64-17-5
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6, 64-17-5
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

Suitable for flash and normal phase chromatography

  • A green alternative to chlorinated solvents such as dichloromethane (DCM)
  • Suitable for use for flash and normal phase chromatography
Absorbance 0.75 AU Maximum(255 nm), 0.35 AU Maximum(260 nm), 0.1 AU Maximum(270 nm), 0.05 AU Maximum(275 nm), 0.03 AU Maximum(300 nm), 0.01 AU Maximum(325 nm), 0.01 AU Maximum(400 nm)
Physical Form Liquid
Grade OPTIMA™
Ethanol 24-26%
Packaging Amber Soda Lime Glass
Residue after Evaporation 10 ppm Maximum
Water 100 ppm Maximum
Color Colorless
Quantity 4L
Chemical Name or Material Ethyl Acetate Ethanol Solution (3:1)

storageNote1 : Flammable