Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Exceeds A.C.S. Specifications, SpectroSolv (TM), 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
Packaging:
Amber Glass Bottle
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O

SP603, 141-78-6, C4H8O2

Spectrum™ Ethyl Acetate, Exceeds A.C.S. Specifications, SpectroSolv (TM) is a colorless liquid that smells like pear drops. Abbreviated as EA, it can be found in glues, nail polish removers, cigarettes and other products. Primarily used as a diluent and solvent thanks in part to its low cost and toxicity it is also used in Laboratory settings for column chromatography and extractions. This SpectroSolv grade product is suitable for ultraviolet spectrophotometry usage and exceeds the specification of an ACS grade quality reagent. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 1
UN Number UN1173
Percent Purity 99.5%
Packaging Amber Glass Bottle
Residue after Evaporation 3e-005
Titratable Acid 0.0009 meq/g
Water 0.002
Quantity 4 x 4 L