Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Formula Weight 88.11
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.90kg
Flash Point −4.00°C (−39.2°F)
Quantity 4L
Chemical Name or Material Ethyl Acetate
Grade HPLC Grade