Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, NF, 98-102%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O
141-78-6
C4H8O2
88.11
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
CCOC(C)=O

ET105, 141-78-6, C4H8O2

Spectrum™ Ethyl Acetate, NF is used as a diluent and solvent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Acetaldehyde 0.001
UN Number UN1173
Percent Purity 98 to 102%
Non_volatile Matter 0.0002
Packaging Amber Glass Bottle
Specific Gravity at 25 degC 0.894-0.898
Quantity 6 x 500 mL