Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate (NF), Fisher Chemical

Click to view available options
Quantity:
4L
20L
Packaging:
Glass Bottle
Steel Pail
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate,acetoxyethane,acetic acid ethyl ester,acetic ether,ethyl acetic ester,vinegar naphtha,acetic acid, ethyl ester,ethylacetate,acetidin,acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
Acidity Pass Test (NF)
Additional Information Vapor Pressure: 103mbar at 20°C
Assay Percent Range 99.0 to 100.5 %
Linear Formula CH3COOC2H5
UN Number 1173
Formula Weight 88.11
Physical Form Liquid
Carbonizable Substances Pass Test
Percent Purity 99.0 to 100.5% (NF)
Grade BP/EP/FCC/JP/NF/USP
Density 0.902g/cm³
Identification Pass Test
Methyl Compounds Pass Test
Non_volatile Matter 0.02% max.
Packaging Glass Bottle
Purity 99.5% min.
Residual Solvents Meets Requirements (USP)
Specific Gravity 0.894 to 0.898 (NF)
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
Boiling Point 75°C
Color Colorless
Melting Point -83.5°C
Quantity 4L
Chemical Name or Material Ethyl Acetate
Show More Show Less

DANGER!

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid