Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6
C4H8O2
88.106
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
For UV/VIS spectroscopy
Absorbance 0.01 max. at 330nm, 0.01 max. at 400nm, 0.05 max. at 275nm, 0.1 max. at 263nm, 0.5 max. at 257nm, 1 max. at 255nm
Assay Percent Range 99.5%
Linear Formula CH3CO2C2H5
Beilstein 02, 125
Merck Index 15, 3811
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Physical Form Liquid
Percent Purity 99.5%
Grade ACS
Density 0.9020g/mL
Packaging EcoSafPak™ Glass Bottle
Refractive Index 1.3710 to 1.3740
Residue after Evaporation 0.003% max.
Specific Gravity 0.902
Substances Darkened by H2SO4 Passes Test
Titratable Acid 0.0009 meq/g max.
Water 0.2% max. (K.F.)
Flash Point −4°C
Viscosity 0.45 mPa.s (20°C)
Boiling Point 75.0°C to 78.0°C
Color Undesignated
Melting Point -83.5°C
Quantity 500mL
Chemical Name or Material Ethyl acetate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-500-4

RTECSNumber : AH5425000

TSCA : TSCA