Ethyl Acetate, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 330WORLDDM55M

Packaging Metal drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-419

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-78-6
    Physical Form Liquid
    Molecular Formula C4H8O2
    Formula Weight 88.1
    Synonym Acetic Acid Ethyl Ester
    Chemical Name or Material Ethyl Acetate

    Specifications

    Formula Weight 88.1
    Quantity 55 gal.
    Color Clear colorless
    UN Number 1173
    Physical Form Liquid
    Odor Characteristic
    Stability Stable under recommended storage conditions.
    CAS 141-78-6
    Description General solvent use
    Packaging Metal drum
    Molecular Formula C4H8O2
    Synonym Acetic Acid Ethyl Ester
    Chemical Name or Material Ethyl Acetate

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit