Ethyl Acetate, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 330WORLDCS4L

Packaging Amber glass bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-101-418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-78-6
    Physical Form Liquid
    Molecular Formula C4H8O2
    Formula Weight 88.1
    Synonym Acetic Acid Ethyl Ester
    Chemical Name or Material Ethyl Acetate

    Specifications

    Boiling Point 75° to 78°C
    Formula Weight 88.1
    Quantity 4L
    Color Clear colorless
    Flash Point −4°C (25°F)
    UN Number 1173
    Physical Form Liquid
    Odor Characteristic
    Packaging Amber glass bottle
    Synonym Acetic Acid Ethyl Ester
    Description General solvent use
    Stability Stable under recommended storage conditions.
    Molecular Formula C4H8O2
    CAS 141-78-6
    Chemical Name or Material Ethyl Acetate

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit