Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl acetimidate hydrochloride, 97%

Click to view available options
Quantity:
10g
50g
250g
2208-07-3
C12H9NO3
215.21
MFCD00012572
VNHGETRQQSYUGZ-UHFFFAOYSA-N
2-nitrodiphenyl ether,2-nitrophenyl phenyl ether,benzene, 1-nitro-2-phenoxy,o-nitrophenyl phenyl ether,2-nde,phenyl o-nitrophenyl ether,ether, o-nitrophenyl phenyl,ether, 2-nitrophenyl phenyl,ether, 2-nitrodiphenyl,,ganoderic-acid-g
16661
1-nitro-2-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2[N+](=O)[O-]
2208-07-3
C12H9NO3
215.21
MFCD00012572
VNHGETRQQSYUGZ-UHFFFAOYSA-N
2-nitrodiphenyl ether,2-nitrophenyl phenyl ether,benzene, 1-nitro-2-phenoxy,o-nitrophenyl phenyl ether,2-nde,phenyl o-nitrophenyl ether,ether, o-nitrophenyl phenyl,ether, 2-nitrophenyl phenyl,ether, 2-nitrodiphenyl,,ganoderic-acid-g
16661
1-nitro-2-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2[N+](=O)[O-]
Applications
Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

Solubility
Soluble in water (50mg/L).

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Sensitivity Moisture sensitive
Melting Point ~108°C (decomposition)
Quantity 250g
Beilstein 3552401
Solubility Information Soluble in water (50mg/L).
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material Ethyl acetimidate hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-631-7

TSCA : Yes

Recommended Storage : Ambient temperatures