Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetoacetate 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Plastic drum
141-97-9
C6H10O3
130.14
MFCD00009199
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl acetoacetate,ethyl acetylacetate,ethyl 3-oxobutyrate,diacetic ether,butanoic acid, 3-oxo-, ethyl ester,ethyl acetylacetonate,acetoacetic acid, ethyl ester,3-oxobutanoic acid ethyl ester,3-oxo-butyric acid ethyl ester,active acetylacetate
8868
CHEBI:4893
ethyl 3-oxobutanoate
CCOC(=O)CC(=O)C
141-97-9
C6H10O3
130.14
MFCD00009199
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl acetoacetate,ethyl acetylacetate,ethyl 3-oxobutyrate,diacetic ether,butanoic acid, 3-oxo-, ethyl ester,ethyl acetylacetonate,acetoacetic acid, ethyl ester,3-oxobutanoic acid ethyl ester,3-oxo-butyric acid ethyl ester,active acetylacetate
8868
CHEBI:4893
ethyl 3-oxobutanoate
CCOC(=O)CC(=O)C
Viscosity 1.696 mPa.s (20°C)
Packaging Plastic drum
Quantity 25L
Infrared Spectrum Authentic
Density 1.0300g/mL
Color Colorless
Water 0.1% max. (K.F.)
Specific Gravity 1.03
Refractive Index 1.4180 to 1.4200
Flash Point 70°C
Boiling Point 180.0°C
Melting Point -45.0°C
Assay Percent Range 99%
Linear Formula CH3COCH2CO2C2H5
Beilstein 03, 632
Fieser 12,222
Merck Index 15, 3812
Solubility Information Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone
Formula Weight 130.14
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material Ethyl acetoacetate, 99%
Show More Show Less

EINECSNumber : 205-516-1

RTECSNumber : AK5250000

TSCA : TSCA