Ethyl Acetoacetate, ≥99%, Alfa Aesar™

Manufacturer: Alfa Aesar A1254436

Quantity 500g

 View more versions of this product

Catalog No. AAA1254436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-97-9
    Physical Form Liquid
    Assay Percent Range ≥99%
    Molecular Formula CH3COCH2CO2CH2CH3
    Formula Weight 130.14
    Chemical Name or Material Ethyl Acetoacetate

    Specifications

    Formula Weight 130.14
    Quantity 500g
    TSCA TSCA
    MDL Number MFCD00009199
    Beilstein 385838
    Merck Index 14,3758
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319-H227
    Causes serious eye irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P337+P313-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cauti
    RTECS Number AK5250000
    Physical Form Liquid
    Density 1.025
    Assay Percent Range ≥99%
    Molecular Formula CH3COCH2CO2CH2CH3
    CAS 141-97-9
    Chemical Name or Material Ethyl Acetoacetate
    Assay ≥99%

    Description

    Applications
    Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

    Solubility
    Miscible with water, benzene, chloroform, ether and acetone.

    Notes
    Incompatible with acids, bases, oxidizing agents, alkali metals and reducing agents.

    GHS H Statement
    H319-H227
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P210-P280-P305+P351+P338-P337+P313-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cauti

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit