Alfa Aesar™ Ethyl adamantane-1-carboxylate, 98+%

Manufacturer: Alfa Aesar™ B2451809

Quantity 10g

 View more versions of this product

Catalog No. AAB2451809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2094-73-7
    Molecular Formula C13H20O2
    Formula Weight 208.3
    MDL Number MFCD00078272
    Beilstein 2050027
    Synonym Adamantane-1-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl adamantane-1-carboxylate

    Specifications

    Formula Weight 208.3
    Molecular Formula C13H20O2
    MDL Number MFCD00078272
    CAS 2094-73-7
    Beilstein 2050027
    Synonym Adamantane-1-carboxylic acid ethyl ester
    Boiling Point 90° to 91°C (1mmHg)
    Quantity 10g
    Chemical Name or Material Ethyl adamantane-1-carboxylate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit