Learn More
Alfa Aesar™ Ethyl adamantane-1-carboxylate, 98+%
Manufacturer:  Alfa Aesar™ B2451804
Quantity 2g
 View more versions of this product
Catalog No. AAB2451804
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
2094-73-7
208.3
2050027
Ethyl adamantane-1-carboxylate
C13H20O2
MFCD00078272
Adamantane-1-carboxylic acid ethyl ester
Specifications
2094-73-7
208.3
2050027
90° to 91°C (1mmHg)
Ethyl adamantane-1-carboxylate
C13H20O2
MFCD00078272
Adamantane-1-carboxylic acid ethyl ester
2g
≥98%