Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 190 Proof, FCC, 94.9%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

E1020, 64-17-5, C2H5OH

Spectrum™ Ethyl Alcohol, 190 Proof, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Acidity 3e-005
Alkalinity 3 mg/kg
UN Number UN1170
Percent Purity 94.9%
Lead (Pb) 0.5 mg/kg
Non_volatile Matter 3e-005
Packaging Amber Glass Bottle
Quantity 500 mL