Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 190 Proof, Reagent, ACS, 95%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Drum
Glass Bottle
Plastic Bottle
64-17-5,7732-18-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5,7732-18-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
CAS Max % 0.95,0.05
Appearance Clear
Physical Form Liquid
Packaging Glass Bottle
Specific Gravity 0.8 at 20°C
Flash Point 17°C (63°F)
Boiling Point 79°C (174.2°F)
Color Colorless
Quantity 4 x 4 L