Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, Completely Denatured, Macron Fine Chemicals™

Click to view available options
Quantity:
20L
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
64-17-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
Quantity 20L
Color Colorless
Physical Form Liquid
Chemical Name or Material Ethyl Alcohol