Ethyl Alcohol Denatured (Proprietary Solvent), Fisher Chemical

Formulation: Ethyl Alcohol (100 parts); Ethyl Acetate (1 part); Methyl iso-Butyl Ketone (1 part); Aviation Gasoline (1 part)

Packaging Glass Bottle
Quantity 500mL

 View more versions of this product

Catalog No. A407-500

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 64-17-5
    MDL Number MFCD00003568
    Synonym Ethanol, Denatured
    Chemical Name or Material Ethyl Alcohol Denatured, Proprietary Solvent

    Specifications

    Boiling Point 78.5°C
    Melting Point −90.0°C
    Quantity 500mL
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Clear Colorless
    Density 0.7905g/cm³
    Specific Gravity 0.7905
    Flash Point 13.9°C (57°F)
    UN Number 1170
    Solubility Information Soluble in water
    Description Formulation: Ethyl Alcohol (100 parts); Ethyl Acetate (1 part); Methyl iso-Butyl Ketone (1 part); Aviation Gasoline (1 part)
    Vapor Pressure 40.9mmHg at 20°C
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Exposure through inhalation may result in delayed pulmonary edema, which may be fatal. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risks of irreversible effects. Causes respiratory tract burns. Irritating to eyes and skin. May cause central nervous system effects. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Oxidizing agent.

    NFPA

    Health:2
    Flammability:3
    Instability:0
    Identification Pass Test
    Additional Information Vapor Pressure: 40.9mmHg at 20°C
    Packaging Glass Bottle
    Cleanup Symbol Solvent
    MDL Number MFCD00003568
    CAS 64-17-5
    ChemAlert Storage Symbol Red
    Synonym Ethanol, Denatured
    Health Hazard 2 DANGER!
    Denaturants Pass Test
    Chemical Name or Material Ethyl Alcohol Denatured, Proprietary Solvent
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Exposure through inhalation may result in delayed pulmonary edema, which may be fatal. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risks of irreversible effects. Causes respiratory tract burns. Irritating to eyes and skin. May cause central nervous system effects. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Oxidizing agent.

    NFPA

    Health:2
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit