Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol Denatured (Proprietary Solvent), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Formulation: Ethyl Alcohol (100 parts); Ethyl Acetate (1 part); Methyl iso-Butyl Ketone (1 part); Aviation Gasoline (1 part)

Packaging Aluminum Can
Quantity 4L

 View more versions of this product

Catalog No. A407S-4



 

Due to product restrictions, please to purchase or view availability for this product.

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

64-17-5
MFCD00003568
Ethanol, Denatured
Ethyl Alcohol Denatured, Proprietary Solvent

Specifications

64-17-5
Liquid
1170
Soluble in water
0.7905g/cm³
Pass Test
0.7905
13.9°C (57°F)
Clear Colorless
4L
Vapor Pressure: 40.9mmHg at 20°C
MFCD00003568
Ethanol, Denatured
Formulation: Ethyl Alcohol (100 parts); Ethyl Acetate (1 part); Methyl iso-Butyl Ketone (1 part); Aviation Gasoline (1 part)
Pass Test
Aluminum Can
40.9mmHg at 20°C
78.5°C
−90.0°C
Ethyl Alcohol Denatured, Proprietary Solvent

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Exposure through inhalation may result in delayed pulmonary edema, which may be fatal. Aspiration hazard if swallowed - can enter lungs and cause damage. Possible risks of irreversible effects. Causes respiratory tract burns. Irritating to eyes and skin. May cause central nervous system effects. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Oxidizing agent.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit