Alfa Aesar™ (Ethyl benzoate)tricarbonylchromium, 98%

Hydrogenation catalyst

Manufacturer: Alfa Aesar™ 4428306

Quantity 5g

 View more versions of this product

Catalog No. AA4428306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32874-26-3
    Molecular Formula C12H10CrO5
    Formula Weight 286.2
    MDL Number MFCD02683559
    UN Number UN3466
    Synonym (Ethyl benzoate)chromium tricarbonyl
    Chemical Name or Material (Ethyl benzoate)tricarbonylchromium

    Specifications

    Formula Weight 286.2
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD02683559
    Synonym (Ethyl benzoate)chromium tricarbonyl
    Health Hazard 1 Danger
    Color Orange
    Quantity 5g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Physical Form Powder
    UN Number UN3466
    Health Hazard 2 GHS H Statement
    H301-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 54° to 59°C
    Sensitivity Air & Moisture Sensitive
    Molecular Formula C12H10CrO5
    CAS 32874-26-3
    Chemical Name or Material (Ethyl benzoate)tricarbonylchromium
    Assay 98%

    GHS H Statement
    H301-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit