Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethyl benzoate)tricarbonylchromium, 98%, Thermo Scientific™

Catalog Number p-7059264
Click to view available options
Quantity:
1 g
32874-26-3
C12H10CrO5
286.203
MFCD02683559
SWSAJUVURPSGIG-UHFFFAOYSA-N
ethyl benzoate tricarbonylchromium, ethyl benzoate tricarbonylchromium 0, tris carbon monoxide ethyl benzoate chromium
13222494
carbon monoxide;chromium;ethyl benzoate
CCOC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]
32874-26-3
C12H10CrO5
286.203
MFCD02683559
SWSAJUVURPSGIG-UHFFFAOYSA-N
ethyl benzoate tricarbonylchromium, ethyl benzoate tricarbonylchromium 0, tris carbon monoxide ethyl benzoate chromium
13222494
carbon monoxide;chromium;ethyl benzoate
CCOC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Hydrogenation catalyst

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 54°C to 59°C
Color Orange
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Formula Weight 286.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Ethyl benzoate)tricarbonylchromium
Hazard Category H301-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H331-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit