Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Caprylate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
106-32-1
C10H20O2
172.27
MFCD00009552
YYZUSRORWSJGET-UHFFFAOYSA-N
ethyl caprylate,octanoic acid, ethyl ester,ethyl n-octanoate,caprylic acid ethyl ester,ethyl octoate,ethyl octylate,octanoic acid ethyl ester,natural,unii-81c5mop582,fema no. 2449
7799
CHEBI:87426
ethyl octanoate
CCCCCCCC(=O)OCC
106-32-1
C10H20O2
172.27
MFCD00009552
YYZUSRORWSJGET-UHFFFAOYSA-N
ethyl caprylate,octanoic acid, ethyl ester,ethyl n-octanoate,caprylic acid ethyl ester,ethyl octoate,ethyl octylate,octanoic acid ethyl ester,natural,unii-81c5mop582,fema no. 2449
7799
CHEBI:87426
ethyl octanoate
CCCCCCCC(=O)OCC
Density 0.8680g/mL
Specific Gravity 0.868
Refractive Index 1.4150 to 1.42
Melting Point -47°C
Flash Point 75°C
Boiling Point 206°C to 208°C
Water 0.3% max. (K.F.)
Packaging Glass bottle
Acidity 1mg KOH/g max.
Color Colorless
Infrared Spectrum Authentic
Quantity 100mL
Assay Percent Range 99+%
Linear Formula CH3(CH2)6CO2C2H5
Beilstein 02, 348
Merck Index 15, 3833
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether,insoluble in glycerol
Formula Weight 172.27
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material Ethyl caprylate, 99+%
Show More Show Less

EINECSNumber : 203-385-5

RTECSNumber : RH0680000

TSCA : TSCA