Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cellulose, FCC, 44-50%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
9004-57-3
C23H24N6O4
448.48
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
9004-57-3
C23H24N6O4
448.48
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

E1036, 9004-57-3

Spectrum™ Ethyl Cellulose, FCC , a derivative of cellulose, is often used as an emulsifier food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.005
Percent Purity 44 to 50%
Lead (Pb) 3 mg/kg
Loss on Drying 0.03
Packaging Poly Bottle
Quantity 500 g