Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Chloride, USP, Spectrum™

Click to view available options
Quantity:
3.5 oz.
Packaging:
Amber Glass Bottle

ET109, 75-00-3, C2H5Cl

Spectrum™ Ethyl Chloride, USP acts as a topical local analgesic when sprayed onto the skin. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS 75-00-3
CAS Max % 1
Molecular Formula C2H5Cl
UN Number UN1037
InChI Key HRYZWHHZPQKTII-UHFFFAOYSA-N
SMILES CCCl
IUPAC Name chloroethane
Molecular Weight (g/mol) 64.51
Packaging Amber Glass Bottle
Quantity 3.5 oz.