Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl chlorofluoroacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
401-56-9
C4H6ClFO2
140.538
MFCD00039333
WUHVJSONZHSDFC-UHFFFAOYSA-N
ethyl chlorofluoroacetate,acetic acid, chlorofluoro-, ethyl ester,chlorofluoroacetic acid ethyl ester,ethyl chloro fluoro acetate,ethyl fluorochloroacetate,acetic acid, 2-chloro-2-fluoro-, ethyl ester,acmc-1csjg,ksc492m6l,ethyl 2-chloro-2-fluoro-acetate
101668
ethyl 2-chloro-2-fluoroacetate
CCOC(=O)C(F)Cl
401-56-9
C4H6ClFO2
140.538
MFCD00039333
WUHVJSONZHSDFC-UHFFFAOYSA-N
ethyl chlorofluoroacetate,acetic acid, chlorofluoro-, ethyl ester,chlorofluoroacetic acid ethyl ester,ethyl chloro fluoro acetate,ethyl fluorochloroacetate,acetic acid, 2-chloro-2-fluoro-, ethyl ester,acmc-1csjg,ksc492m6l,ethyl 2-chloro-2-fluoro-acetate
101668
ethyl 2-chloro-2-fluoroacetate
CCOC(=O)C(F)Cl
Quantity 5g
Density 1.2250g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Refractive Index 1.3940 to 1.3960 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula FCH(Cl)COOC2H5
Formula Weight 140.54
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl chlorofluoroacetate, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger