Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cis-2-isothiocyanato-1-cyclopentanecarboxylate, 97%, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass Bottle in Plastic Container
311341-94-3
C9H13NO2S
199.268
MFCD01863282
IDCQKRRLLDGLAD-SFYZADRCSA-N
ethyl 1r,2s-2-isothiocyanatocyclopentane-1-carboxylate,ethyl 2s,1r-2-isothiocyanatocyclopentanecarboxylate,2beta-isothiocyanatocyclopentane-1beta-carboxylic acid ethyl ester,ethyl cis-2-isothiocyanato-1-cyclopentanecarboxylate, 97,cyclopentanecarboxylicacid, 2-isothiocyanato-, ethyl ester, 1r,2s-rel,cyclopentanecarboxylic acid, 2-isothiocyanato-, ethyl ester, 1r,2s-rel-9ci
2733737
ethyl (1R,2S)-2-isothiocyanatocyclopentane-1-carboxylate
CCOC(=O)C1CCCC1N=C=S
311341-94-3
C9H13NO2S
199.268
MFCD01863282
IDCQKRRLLDGLAD-SFYZADRCSA-N
ethyl 1r,2s-2-isothiocyanatocyclopentane-1-carboxylate,ethyl 2s,1r-2-isothiocyanatocyclopentanecarboxylate,2beta-isothiocyanatocyclopentane-1beta-carboxylic acid ethyl ester,ethyl cis-2-isothiocyanato-1-cyclopentanecarboxylate, 97,cyclopentanecarboxylicacid, 2-isothiocyanato-, ethyl ester, 1r,2s-rel,cyclopentanecarboxylic acid, 2-isothiocyanato-, ethyl ester, 1r,2s-rel-9ci
2733737
ethyl (1R,2S)-2-isothiocyanatocyclopentane-1-carboxylate
CCOC(=O)C1CCCC1N=C=S
Packaging Glass Bottle in Plastic Container
Infrared Spectrum Authentic
Quantity 500mg
Color Yellow
Refractive Index 1.5125 to 1.5165 (20°C, 589nm)
Assay Percent Range 97%
Linear Formula C5H8(NCS)COOCH2CH3
Formula Weight 199.3
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Ethyl cis-2-isothiocyanato-1-cyclopentanecarboxylate, 97%
Show More Show Less

MOISTURE SENSITIVE

STENCH