Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cyanoacetate, 98+%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
105-56-6
C5H7NO2
113.12
ZIUSEGSNTOUIPT-UHFFFAOYSA-N
ethyl cyanoacetate,ethyl cyanacetate,cyanoacetic acid ethyl ester,acetic acid, cyano-, ethyl ester,ethyl cyanoethanoate,cyanoacetic ester,cyanacetate ethyle,usaf kf-25,estere cianoacetico,ethyl cyano acetate
7764
ethyl 2-cyanoacetate
CCOC(=O)CC#N
105-56-6
C5H7NO2
113.12
ZIUSEGSNTOUIPT-UHFFFAOYSA-N
ethyl cyanoacetate,ethyl cyanacetate,cyanoacetic acid ethyl ester,acetic acid, cyano-, ethyl ester,ethyl cyanoethanoate,cyanoacetic ester,cyanacetate ethyle,usaf kf-25,estere cianoacetico,ethyl cyano acetate
7764
ethyl 2-cyanoacetate
CCOC(=O)CC#N
Viscosity 1.01 mPa.s (20°C)
Density 1.0600g/mL
Quantity 1kg
Refractive Index 1.4170 to 1.4190
Color Undesignated
Boiling Point 208.0°C to 210.0°C
Flash Point 110°C
Packaging Glass bottle
Specific Gravity 1.06
Water 0.1% max. (K.F.)
Melting Point -22.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula NCCH2CO2C2H5
Beilstein 02,585
Fieser 13,69
Merck Index 15,3841
Solubility Information Solubility in water: 20g/L(20°C). Other solubilities: soluble in ammonium hydroxide,diethyl ether,,ethanol,aqueous solutions of alkalies
Formula Weight 113.12
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Ethyl cyanoacetate
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.