Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cyanoformate 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
623-49-4
C4H5NO2
99.09
MFCD00001836
MSMGXWFHBSCQFB-UHFFFAOYSA-N
carbonocyanidic acid, ethyl ester,ethyl carbonocyanidate,2-ethoxy-2-oxoacetonitrile,cyanoformic acid ethyl ester,ethyl carbonocyanidoate,formic acid, cyano-, ethyl ester,cyanocarbonyloxyethane,ethylcyanidocarbonat,ethyl cyanoformate,ethyl cyanocarbonate
69336
ethyl cyanoformate
CCOC(=O)C#N
623-49-4
C4H5NO2
99.09
MFCD00001836
MSMGXWFHBSCQFB-UHFFFAOYSA-N
carbonocyanidic acid, ethyl ester,ethyl carbonocyanidate,2-ethoxy-2-oxoacetonitrile,cyanoformic acid ethyl ester,ethyl carbonocyanidoate,formic acid, cyano-, ethyl ester,cyanocarbonyloxyethane,ethylcyanidocarbonat,ethyl cyanoformate,ethyl cyanocarbonate
69336
ethyl cyanoformate
CCOC(=O)C#N
Flash Point 24°C
Boiling Point 115°C to 116°C
Specific Gravity 1
Refractive Index 1.3810 to 1.383
Density 1.0000g/mL
Quantity 50g
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula NCCO2C2H5
Beilstein 02, 547
Fieser 07,145
Formula Weight 99.09
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl cyanoformate, 99%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-797-9

TSCA : TSCA