Alfa Aesar™ Ethyl cycloheptylacetate, 98%

Manufacturer: Alfa Aesar™ H2586606

Quantity 5g

 View more versions of this product

Catalog No. AAH2586606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80246-70-4
    Molecular Formula C11H20O2
    Formula Weight 184.28
    MDL Number MFCD08437072
    Synonym Cycloheptylacetic acid ethyl ester; Ethyl cycloheptaneacetate
    Chemical Name or Material Ethyl cycloheptylacetate

    Specifications

    Formula Weight 184.28
    Boiling Point 80° to 85°C (2mmHg)
    Molecular Formula C11H20O2
    MDL Number MFCD08437072
    CAS 80246-70-4
    Synonym Cycloheptylacetic acid ethyl ester; Ethyl cycloheptaneacetate
    Quantity 5g
    Chemical Name or Material Ethyl cycloheptylacetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit