Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cyclohexylacetate, 98+%

Click to view available options
Quantity:
5g
5452-75-5
C10H18O2
170.252
ZBDAMDWKXGTKBT-UHFFFAOYSA-N
ethyl cyclohexylacetate,ethyl cyclohexaneacetate,cyclohexaneacetic acid, ethyl ester,ethyl-2-cyclohexylacetate,ethylcyclohexylacetate,unii-v36o028g9r,cyclohexylacetic acid ethyl ester,cyclohexaneacetic acid ethyl ester,cyclohexyl acetic acid ethyl ester,pubchem20245
21595
ethyl 2-cyclohexylacetate
CCOC(=O)CC1CCCCC1
5452-75-5
C10H18O2
170.252
ZBDAMDWKXGTKBT-UHFFFAOYSA-N
ethyl cyclohexylacetate,ethyl cyclohexaneacetate,cyclohexaneacetic acid, ethyl ester,ethyl-2-cyclohexylacetate,ethylcyclohexylacetate,unii-v36o028g9r,cyclohexylacetic acid ethyl ester,cyclohexaneacetic acid ethyl ester,cyclohexyl acetic acid ethyl ester,pubchem20245
21595
ethyl 2-cyclohexylacetate
CCOC(=O)CC1CCCCC1
Quantity 5g
Flash Point 80°C (176°F)
Boiling Point 211°C to 212°C
Formula Weight 170.25
Chemical Name or Material Ethyl cyclohexylacetate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA