Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl cyclopropanecarboxylate, 98%

Click to view available options
Quantity:
25g
100g
500g
4606-07-9
C6H10O2
114.14
MFCD00001282
LDDOSDVZPSGLFZ-UHFFFAOYSA-N
cyclopropanecarboxylic acid ethyl ester,ethyl cyclopropylcarboxylate,ethyl cyclopropane carboxylate,cyclopropanecarboxylic acid, ethyl ester,pubchem11840,ethyl cylopropylcarboxylate,ethoxycarbonyl cyclopropane,ethylcyclopropane carboxylate,ksc497s5p,lddosdvzpsglfz-uhfffaoysa
78351
ethyl cyclopropanecarboxylate
CCOC(=O)C1CC1
4606-07-9
C6H10O2
114.14
MFCD00001282
LDDOSDVZPSGLFZ-UHFFFAOYSA-N
cyclopropanecarboxylic acid ethyl ester,ethyl cyclopropylcarboxylate,ethyl cyclopropane carboxylate,cyclopropanecarboxylic acid, ethyl ester,pubchem11840,ethyl cylopropylcarboxylate,ethoxycarbonyl cyclopropane,ethylcyclopropane carboxylate,ksc497s5p,lddosdvzpsglfz-uhfffaoysa
78351
ethyl cyclopropanecarboxylate
CCOC(=O)C1CC1
Applications
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Boiling Point 133°C to 134°C
Flash Point 18°C (64°F)
Quantity 25g
UN Number UN3272
Beilstein 2039752
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material Ethyl cyclopropanecarboxylate

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.