Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl dichlorophosphate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1498-51-7
C2H5Cl2O2P
162.94
MFCD00002069
YZBOZNXACBQJHI-UHFFFAOYSA-N
ethyl dichlorophosphate,ethyl phosphorodichloridate,phosphorodichloridic acid, ethyl ester,ethyldichlorophosphate,dichloroethoxyphosphine oxide,ethylphosphoric acid dichloride,dichlorophosphoric acid, ethyl ester,unii-4ip5dtw75u,ethylesterdichlorid german,hsdb 417
15158
1-dichlorophosphoryloxyethane
CCOP(=O)(Cl)Cl
1498-51-7
C2H5Cl2O2P
162.94
MFCD00002069
YZBOZNXACBQJHI-UHFFFAOYSA-N
ethyl dichlorophosphate,ethyl phosphorodichloridate,phosphorodichloridic acid, ethyl ester,ethyldichlorophosphate,dichloroethoxyphosphine oxide,ethylphosphoric acid dichloride,dichlorophosphoric acid, ethyl ester,unii-4ip5dtw75u,ethylesterdichlorid german,hsdb 417
15158
1-dichlorophosphoryloxyethane
CCOP(=O)(Cl)Cl
Boiling Point 167°C (758.0mmHg)
Flash Point >110°C
Specific Gravity 1.373
Refractive Index 1.4330 to 1.435
Quantity 100g
Color Brown to Colorless
Density 1.3730g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C2H5OP(O)Cl2
Beilstein 01,332
Formula Weight 162.94
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Ethyl dichlorophosphate, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed. Causes severe skin burns and eye damage. Fatal if inhaled. Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Wash face, hands and any exposed skin thoroughly after handling.

Danger

EINECSNumber : 216-099-0

RTECSNumber : TD4390000

TSCA : TSCA