Alfa Aesar™ Ethyl diethoxyphosphinylformate, 98%

Manufacturer: Alfa Aesar™ B2336736

Quantity 500g

 View more versions of this product

Catalog No. AAB2336736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1474-78-8
    Molecular Formula C7H15O5P
    Formula Weight 210.17
    MDL Number MFCD00009108
    Beilstein 1781949
    Synonym Triethyl phosphonoformate; Diethoxyphosphinylformic acid ethyl ester
    Chemical Name or Material Ethyl diethoxyphosphinylformate

    Specifications

    Formula Weight 210.17
    Molecular Formula C7H15O5P
    Synonym Triethyl phosphonoformate; Diethoxyphosphinylformic acid ethyl ester
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 119°C (246°F)
    TSCA TSCA
    MDL Number MFCD00009108
    CAS 1474-78-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Boiling Point 128° to 129°C (9mmHg)
    Quantity 500g
    Beilstein 1781949
    Chemical Name or Material Ethyl diethoxyphosphinylformate
    Assay 98%

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit