Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl diethoxyphosphinylformate, 98%

Click to view available options
Quantity:
25g
100g
500g
1474-78-8
C7H15O5P
210.166
MFCD00009108
NOJFJZZMRDSOLM-UHFFFAOYSA-N
ethyl diethoxyphosphinylformate,triethyl phosphonoformate,triethyl carboxyphosphonate,diethyl ethoxycarbonylphosphonate,triethyl phosphonomethanoate,diethyl carbethoxyphosphonate,ethyldiethoxyphosphinylformate,ethyl diethoxyphosphoryl formate,unii-tux6m928id,phosphinecarboxylic acid, diethoxy-, ethyl ester, oxide
65081
ethyl diethoxyphosphorylformate
CCOC(=O)P(=O)(OCC)OCC
1474-78-8
C7H15O5P
210.166
MFCD00009108
NOJFJZZMRDSOLM-UHFFFAOYSA-N
ethyl diethoxyphosphinylformate,triethyl phosphonoformate,triethyl carboxyphosphonate,diethyl ethoxycarbonylphosphonate,triethyl phosphonomethanoate,diethyl carbethoxyphosphonate,ethyldiethoxyphosphinylformate,ethyl diethoxyphosphoryl formate,unii-tux6m928id,phosphinecarboxylic acid, diethoxy-, ethyl ester, oxide
65081
ethyl diethoxyphosphorylformate
CCOC(=O)P(=O)(OCC)OCC
Boiling Point 128°C to 129°C (9mmHg)
Quantity 100g
Flash Point 119°C (246°F)
Beilstein 1781949
Formula Weight 210.17
Percent Purity 98%
Chemical Name or Material Ethyl diethoxyphosphinylformate

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA