Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl dimethylpyruvate, 97%, Thermo Scientific Chemicals

Catalog Number AAH5341306
Click to view available options
Quantity:
5 g
25 g
20201-24-5
C7H12O3
144.17
MFCD00009122
CKTYYUQUWFEUCO-UHFFFAOYSA-N
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
88406
ethyl 3-methyl-2-oxobutanoate
CCOC(=O)C(=O)C(C)C
20201-24-5
C7H12O3
144.17
MFCD00009122
CKTYYUQUWFEUCO-UHFFFAOYSA-N
ethyl 3-methyl-2-oxobutyrate, ethyl dimethylpyruvate, ethyl 2-oxo-3-methylbutanoate, butanoic acid, 3-methyl-2-oxo-, ethyl ester, acmc-20akzg, ketovaline ethyl ester, pubchem18179, ethyl3-methyl-2-oxobutanoate, ethyl 2-methyl-2-oxobutyrate, ethyl-3-methyl-2-oxobutanoate
88406
ethyl 3-methyl-2-oxobutanoate
CCOC(=O)C(=O)C(C)C

Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.989
Boiling Point 62°C (11 mmHg)
Flash Point 43°C (109°F)
Refractive Index 1.41
Quantity 5 g
UN Number UN3272
Beilstein 1756668
Solubility Information Slightly soluble in water.
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material Ethyl dimethylpyruvate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 243-587-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit