Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl DL-2-hydroxycaproate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
52089-55-1
C8H16O3
160.213
MFCD00004583
MRYSSTRVUMCKKB-UHFFFAOYSA-N
ethyl 2-hydroxycaproate,ethyl dl-2-hydroxycaproate,acmc-20miyg,hexanoic acid, 2-hydroxy-, ethyl ester, r,acmc-20al12,ethyl 2-dl-hydroxyhexanoate,ethyl 2-hydroxyhexanoate,ethyl +/--2-hydroxycaproate,2-hydroxy-hexanoic acid ethyl ester
103589
ethyl 2-hydroxyhexanoate
CCCCC(C(=O)OCC)O
52089-55-1
C8H16O3
160.213
MFCD00004583
MRYSSTRVUMCKKB-UHFFFAOYSA-N
ethyl 2-hydroxycaproate,ethyl dl-2-hydroxycaproate,acmc-20miyg,hexanoic acid, 2-hydroxy-, ethyl ester, r,acmc-20al12,ethyl 2-dl-hydroxyhexanoate,ethyl 2-hydroxyhexanoate,ethyl +/--2-hydroxycaproate,2-hydroxy-hexanoic acid ethyl ester
103589
ethyl 2-hydroxyhexanoate
CCCCC(C(=O)OCC)O
Quantity 10g
Packaging Glass bottle
Density 0.9600g/mL
Assay Percent Range 97%
Linear Formula CH3(CH2)3CH(OH)CO2C2H5
Beilstein 03, 333
Formula Weight 160.21
Chemical Name or Material Ethyl DL-2-hydroxycaproate, 97%