Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, ACS, 98%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Drum
Steel Can
Steel Pail
60-29-7
60-29-7
CAS Max % 1
Percent Purity 0.98
Grade ACS
Packaging Steel Can
Residue after Evaporation 1e-05
Titratable Acid 0.0002 meq/g
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.