Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, GR, ACS, 98.0%, MilliporeSigma™

Catalog No. p-7162450
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Aluminum Drum
Steel Drum
Tinplate Metal Can

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point −116.3°C
Boiling Point 34.6°C (1.013 hPa)
Quantity 1 L
Flash Point −40°C
Packaging Tinplate Metal Can
Residue after Evaporation 0.001% max
Assay Percent Range 98.0% min
Vapor Pressure 587 hPa (20°C)
Solubility Information 69 g/L
Recommended Storage Store at 2 to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.