Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, UniSolv(R), 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
60-29-7
C4H10O
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
CCOCC
60-29-7
C4H10O
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
CCOCC

US621, 60-29-7, C4H10O

Spectrum™ Ethyl Ether, UniSolv(R) is also referred to as sulfuric ether, diethyl ether or even just simply as ether. This liquid was historically used as an anesthetic, but in more recent years been replaced by other more effective anesthetics. It has however some other uses such as fuel and solvent. It′s an organic compound that is extremely flammable and volatile on top of being colorless.
CAS Max % 1
UN Number UN1155
Percent Purity 99.9%
Packaging Amber Glass Bottle
Peroxides 1 ppm
Residue after Evaporation 1 ppm
Titratable Acid 0.0002 meq/g
Water 0.0002
Quantity 4 L