Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (ethoxymethylene)cyanoacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Glass bottle
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-VOTSOKGWSA-N
ethyl ethoxymethylene cyanoacetate,e-ethyl 2-cyano-3-ethoxyacrylate,ethyl 2-cyano-3-ethoxyacrylate,ethyl 3-ethoxy-2-cyanoacrylate,ethyl 2e-2-cyano-3-ethoxyprop-2-enoate,ethyl cyano ethoxymethylene acetate,2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester,2-cyano-3-ethoxy-2-propenoic acid ethyl ester,2-cyano-3-ethoxyacrylic acid ethyl ester,ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
CCOC=C(C#N)C(=O)OCC
94-05-3
C8H11NO3
169.18
MFCD00009136
KTMGNAIGXYODKQ-VOTSOKGWSA-N
ethyl ethoxymethylene cyanoacetate,e-ethyl 2-cyano-3-ethoxyacrylate,ethyl 2-cyano-3-ethoxyacrylate,ethyl 3-ethoxy-2-cyanoacrylate,ethyl 2e-2-cyano-3-ethoxyprop-2-enoate,ethyl cyano ethoxymethylene acetate,2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester,2-cyano-3-ethoxy-2-propenoic acid ethyl ester,2-cyano-3-ethoxyacrylic acid ethyl ester,ethyl 2e-2-cyano-3-ethoxy-2-propenoate
1715183
ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
CCOC=C(C#N)C(=O)OCC
Boiling Point 190°C to 191°C (30.0mmHg)
Flash Point 130°C
Color White to Yellow
Infrared Spectrum Authentic
Quantity 1kg
Packaging Glass bottle
Melting Point 48°C to 52°C
Assay Percent Range 98%
Linear Formula C2H5OCH=C(CN)CO2C2H5
Beilstein 03,47
Fieser 01,38
Solubility Information Solubility in water: <0.01g/L (20°C)
Formula Weight 169.18
Physical Form Crystalline Mass or Crystals
Percent Purity ≥97.5%
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate, 98%
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-299-5

TSCA : TSCA