Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl fluoroacetate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
459-72-3
C4H7FO2
106.1
MFCD00000450
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl fluoroacetate,ethyl monofluoroacetate,acetic acid, fluoro-, ethyl ester,fluoroacetic acid ethyl ester,unii-aus1l6um7b,ethylester kyseliny fluoroctove,monofluoroacetic acid ethyl ester,aus1l6um7b,ethylester kyseliny fluoroctove czech,acetic acid, 2-fluoro-, ethyl ester
9988
ethyl 2-fluoroacetate
CCOC(=O)CF
459-72-3
C4H7FO2
106.1
MFCD00000450
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl fluoroacetate,ethyl monofluoroacetate,acetic acid, fluoro-, ethyl ester,fluoroacetic acid ethyl ester,unii-aus1l6um7b,ethylester kyseliny fluoroctove,monofluoroacetic acid ethyl ester,aus1l6um7b,ethylester kyseliny fluoroctove czech,acetic acid, 2-fluoro-, ethyl ester
9988
ethyl 2-fluoroacetate
CCOC(=O)CF
Refractive Index 1.3745 to 1.3765
Specific Gravity 1.09
Boiling Point 119.2°C (753.0 mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Flash Point 30°C
Quantity 50g
Density 1.0900g/mL
Assay Percent Range 97%
Linear Formula FCH2CO2C2H5
Beilstein 02, 193
Fieser 05,304
Merck Index 15, 4199
Solubility Information Solubility in water: decomposes.
Formula Weight 106.1
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl fluoroacetate, 97%
Show More Show Less

GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-297-8

RTECSNumber : AH7175000

TSCA : TSCA