Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl heptafluorobutyrate, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
356-27-4
C6H5F7O2
242.093
MFCD00000434
JVHJRIQPDBCRRE-UHFFFAOYSA-N
ethyl heptafluorobutyrate,ethyl perfluorobutyrate,heptafluorobutyric acid ethyl ester,butanoic acid, heptafluoro-, ethyl ester,ethyl heptafluorobutanoate,ethyl perfluorobutanoate,ethyl heptafluoro-n-butyrate,butyric acid, heptafluoro-, ethyl ester,butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester,pubchem12539
9644
ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F
356-27-4
C6H5F7O2
242.093
MFCD00000434
JVHJRIQPDBCRRE-UHFFFAOYSA-N
ethyl heptafluorobutyrate,ethyl perfluorobutyrate,heptafluorobutyric acid ethyl ester,butanoic acid, heptafluoro-, ethyl ester,ethyl heptafluorobutanoate,ethyl perfluorobutanoate,ethyl heptafluoro-n-butyrate,butyric acid, heptafluoro-, ethyl ester,butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester,pubchem12539
9644
ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F
Quantity 25g
Packaging Glass bottle
Refractive Index 1.3010 to 1.3030 (20°C, 589nm)
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CF3CF2CF2CO2C2H5
Formula Weight 242.09
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Ethyl heptafluorobutyrate, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-602-1

RTECSNumber : ET4200000

TSCA : TSCA